Gene Transcripts

Gene Symbol
FBXO22
Entrez Gene ID
26263
Gene Name
F-box protein 22
Gene Aliases
FBX22, FISTC1
Chromosome Location
Chr. 15: 75903859 - 75937513 on Build GRCh38
UniGene ID
Hs.591115
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012170.3 NP_036302.1 6 795
NM_147188.2 NP_671717.1 6 795
NR_037623.1 5 656
XM_011521447.2 XP_011519749.1 7 795
GenBank mRNA AK024048.1 - 6 776
AK223500.1 - 6 752
AK294311.1 - 5 579
AK307084.1 - 4 594
AY005144.1 - 6 690
BC020204.1 - 6 795 4546640
BC032540.1 - 6 713 5529506
BC039024.1 - 5 614 6071245
BC041691.1 - 6 762 5246992

View Full Product Details