Gene Transcripts

Gene Symbol
ELP4
Entrez Gene ID
26610
Gene Name
elongator acetyltransferase complex subunit 4
Gene Aliases
AN, C11orf19, PAX6NEB, PAXNEB, dJ68P15A.1, hELP4
Chromosome Location
Chr. 11: 31509729 - 31784525 on Build GRCh38
UniGene ID
Hs.175534
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288725.1 NP_001275654.1 7 933
NM_001288726.1 NP_001275655.1 7 930
NM_019040.4 NP_061913.3 7 930
GenBank mRNA AK000505.1 - 7 909
AK304885.1 - 7 892
AL136677.1 - 7 887
BC012514.1 - 7 881 4509903

View Full Product Details