Gene Transcripts

Gene Symbol
SNX24
Entrez Gene ID
28966
Gene Name
sorting nexin 24
Gene Aliases
PRO1284, SBBI31
Chromosome Location
Chr. 5: 122845465 - 123029318 on Build GRCh38
UniGene ID
Hs.483200
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014035.2 NP_054754.1 4 495
XM_005271972.4 XP_005272029.1 4 384
XM_006714592.3 XP_006714655.1 4 450
XM_011543349.2 XP_011541651.1 5 515
XM_011543350.2 XP_011541652.1 5 514
XM_017009394.1 XP_016864883.1 4 320
GenBank mRNA AF139461.1 - 4 384
AY044655.1 - 4 326
AY358098.1 - 4 344
BC010886.1 - 4 455 3538426
BC069012.1 - 4 442 4585781
BQ278046.1 - 4 467

View Full Product Details