Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127384.2 | NP_001120856.1 | 6 | 814 | |
NM_001291133.1 | NP_001278062.1 | 7 | 891 | ||
NM_013266.3 | NP_037398.2 | 6 | 861 | ||
XM_017016151.1 | XP_016871640.1 | 7 | 856 | ||
XM_017016152.1 | XP_016871641.1 | 8 | 1033 | ||
XM_017016153.1 | XP_016871642.1 | 7 | 864 | ||
XM_017016154.1 | XP_016871643.1 | 2 | 232 | ||
XM_017016155.1 | XP_016871644.1 | 2 | 242 | ||
XM_017016156.1 | XP_016871645.1 | 2 | 119 | ||
XM_017016158.1 | XP_016871647.1 | 7 | 962 | ||
GenBank mRNA | AF091606.1 | - | 6 | 839 | |
AK289756.1 | - | 6 | 750 | ||
AK308726.1 | - | 5 | 561 | ||
AK313882.1 | - | 6 | 864 | ||
BC022004.2 | - | 7 | 886 | ||
BC030646.2 | - | 7 | 886 | ||
BC065819.1 | - | 6 | 757 | 6189390 |