Gene Transcripts

Gene Symbol
TRPM5
Entrez Gene ID
29850
Gene Name
transient receptor potential cation channel subfamily M member 5
Gene Aliases
LTRPC5, MTR1
Chromosome Location
Chr. 11: 2403962 - 2423424 on Build GRCh38
UniGene ID
Hs.272287
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014555.3 NP_055370.1 6 849
XM_017017628.1 XP_016873117.1 7 936
GenBank mRNA AF177473.1 - 6 849
AJ270996.1 - 6 822
BC093787.1 - 6 846 7939632
BC093789.1 - 6 846 7939634
BC143351.1 - 6 846 9051856
BC143353.1 - 6 846 9051858

View Full Product Details