Gene Transcripts

Gene Symbol
GNL2
Entrez Gene ID
29889
Gene Name
G protein nucleolar 2
Gene Aliases
HUMAUANTIG, NGP1, Ngp-1, Nog2, Nug2
Chromosome Location
Chr. 1: 37566812 - 37595985 on Build GRCh38
UniGene ID
Hs.75528
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323623.1 NP_001310552.1 4, 5 535
NM_001323624.1 NP_001310553.1 4, 5 535
NM_013285.2 NP_037417.1 4, 5 535
GenBank mRNA AK307352.1 - 4, 5 489
AK312915.1 - 4, 5 489
BC000107.2 - 4, 5 471 3506131
BC009250.2 - 4, 5 471 2960125
L05425.1 - 4, 5 451

View Full Product Details