Gene Transcripts

Gene Symbol
POMT2
Entrez Gene ID
29954
Gene Name
protein O-mannosyltransferase 2
Gene Aliases
LGMD2N, MDDGA2, MDDGB2, MDDGC2
Chromosome Location
Chr. 14: 77274956 - 77320885 on Build GRCh38
UniGene ID
Hs.132989
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013382.5 NP_037514.2 18 2010
XM_011536675.2 XP_011534977.1 18 2010
XM_011536676.2 XP_011534978.1 17 1923
XM_011536677.2 XP_011534979.1 14 1552
XM_011536679.2 XP_011534981.1 14 1212
XM_017021244.1 XP_016876733.1 17 1844
XM_017021245.1 XP_016876734.1 14 1298
GenBank mRNA AB209114.1 - 18 1832
AF105020.1 - 18 2013
AK023804.1 - 12 2601
AK025962.1 - 8 629
AY090480.1 - 18 2013
BC031651.1 - 18 1900 5169145
BE797202.1 - 415
CR627447.1 - 18 1580
DA056935.1 - 3 167

View Full Product Details