Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016190.2 | NP_057274.1 | 2 | 195 | |
GenBank mRNA | AF077831.1 | - | 2 | 173 | |
AK291301.1 | - | 2 | 184 | ||
AK298640.1 | - | 2 | 184 | ||
AK316568.1 | - | 2 | 184 | ||
BC030807.1 | - | 2 | 184 | 4271624 | |
DA420219.1 | - | 2 | 195 |