Gene Transcripts

Gene Symbol
TAS2R9
Entrez Gene ID
50835
Gene Name
taste 2 receptor member 9
Gene Aliases
T2R9, TRB6
Chromosome Location
Chr. 12: 10809094 - 10810168 on Build GRCh38
UniGene ID
Hs.272391
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023917.2 NP_076406.1 1 143
GenBank mRNA BC069066.1 - 1 50
BC095519.1 - 1 144 6971937

View Full Product Details