Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016060.2 | NP_057144.1 | 4 | 380 | |
XM_017024710.1 | XP_016880199.1 | 3 | 453 | ||
GenBank mRNA | AF151883.1 | - | 4 | 352 | |
AK130946.1 | - | 3 | 283 | ||
AK311875.1 | - | 4 | 349 | ||
BC012539.1 | - | 4 | 355 | 4248123 |