Gene Transcripts

Gene Symbol
SCCPDH
Entrez Gene ID
51097
Gene Name
saccharopine dehydrogenase (putative)
Gene Aliases
CGI-49, NET11
Chromosome Location
Chr. 1: 246724076 - 246768137 on Build GRCh38
UniGene ID
Hs.498397
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016002.2 NP_057086.2 7, 8 1168
GenBank mRNA AF151807.1 - 7, 8 818
AK075178.1 - 7, 8 924
BC026185.2 - 7, 8 829 4798194

View Full Product Details