Gene Transcripts

Gene Symbol
RMDN1
Entrez Gene ID
51115
Gene Name
regulator of microtubule dynamics 1
Gene Aliases
CGI-90, FAM82B, RMD-1, RMD1
Chromosome Location
Chr. 8: 86467116 - 86514348 on Build GRCh38
UniGene ID
Hs.145386
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286707.1 NP_001273636.1 5, 6 704
NM_001286719.1 NP_001273648.1 5, 6 704
NM_001317807.1 NP_001304736.1 6, 7 612
NM_016033.2 NP_057117.2 6, 7 794
XM_005250944.3 XP_005251001.1 6, 7 700
XM_005250947.3 XP_005251004.1 5, 6 610
XM_005250948.3 XP_005251005.1 6, 7 602
XM_006716573.3 XP_006716636.1 6, 7 789
XM_011517090.2 XP_011515392.1 6, 7 774
XM_011517091.2 XP_011515393.1 6, 7 774
XM_011517092.2 XP_011515394.1 5, 6 684
XM_011517093.2 XP_011515395.1 6, 7 774
XM_011517094.2 XP_011515396.1 6, 7 774
XM_011517095.2 XP_011515397.1 6, 7 774
XM_011517096.2 XP_011515398.1 6, 7 774
XM_011517097.2 XP_011515399.1 5, 6 684
XM_011517098.2 XP_011515400.1 5, 6 684
XM_017013518.1 XP_016869007.1 6, 7 700
XM_017013519.1 XP_016869008.1 6, 7 774
XM_017013520.1 XP_016869009.1 6, 7 700
XM_017013521.1 XP_016869010.1 6, 7 700
XM_017013522.1 XP_016869011.1 6, 7 774
XM_017013523.1 XP_016869012.1 6, 7 700
XM_017013524.1 XP_016869013.1 6, 7 602
XM_017013525.1 XP_016869014.1 6, 7 602
XM_017013526.1 XP_016869015.1 6, 7 602
XM_017013527.1 XP_016869016.1 5, 6 699
XM_017013528.1 XP_016869017.1 5, 6 507
GenBank mRNA AF151848.1 - 6, 7 706
AK000672.1 - 6, 7 702
AK297896.1 - 5, 6 605
AK303122.1 - 5, 6 556
AL561197.3 - 6, 7 586
BC008509.1 - 4, 5 358
BC009671.1 - 6, 7 686 3896168

View Full Product Details