Gene Transcripts

Gene Symbol
COQ4
Entrez Gene ID
51117
Gene Name
coenzyme Q4
Gene Aliases
CGI-92, COQ10D7
Chromosome Location
Chr. 9: 128322486 - 128334072 on Build GRCh38
UniGene ID
Hs.98541
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001305942.1 NP_001292871.1 3 636
NM_016035.4 NP_057119.2 4 733
XM_017014792.1 XP_016870281.1 3 658
XM_017014793.1 XP_016870282.1 3 658
GenBank mRNA AF151850.1 - 4 704
AK025376.1 - 4 647
AK301333.1 - 4 402
AK313650.1 - 4 716
BC011895.2 - 4 403 3944239
CB111694.1 - 3 307
CX787359.1 - 4 729
EU216419.1 - 4 715
EU216420.1 - 4 633
EU216421.1 - 4 467
EU216422.1 - 4 405
EU216423.1 - 3 329

View Full Product Details