Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001135643.1 | NP_001129115.1 | 1 | 185 | |
NM_016221.3 | NP_057305.1 | 1 | 185 | ||
XM_005268457.1 | XP_005268514.1 | 1 | 123 | ||
XM_011537646.1 | XP_011535948.1 | 1 | 185 | ||
GenBank mRNA | AF195120.1 | - | 1 | 100 | |
AK000299.1 | - | 1 | 104 | ||
AK296787.1 | - | 1 | 104 | ||
AK307757.1 | - | 1 | 124 | ||
AK311693.1 | - | 1 | 121 | ||
BC026323.1 | - | 1 | 110 | 4815273 | |
DC321382.1 | - | 1 | 185 | ||
KU178637.1 | - | 1 | 83 | ||
KU178638.1 | - | 1 | 83 | ||
KU178639.1 | - | 1 | 83 |