Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016305.3 | NP_057389.1 | 2 | 252 | |
XM_017006548.1 | XP_016862037.1 | 1 | 1264 | ||
XM_017006549.1 | XP_016862038.1 | 2 | 223 | ||
GenBank mRNA | AF201950.1 | - | 1 | 142 | |
AK312225.1 | - | 1 | 47 | ||
BC017804.1 | - | 1 | 70 | 4658643 | |
BC020444.1 | - | 1 | 42 | ||
BX114027.1 | - | 2 | 246 | ||
DR158587.1 | - | 3 | 680 | ||
EL954786.1 | - | 2 | 255 |