Gene Transcripts
- Gene Symbol
- CLEC1B
- Entrez Gene ID
- 51266
- Gene Name
- C-type lectin domain family 1 member B
- Gene Aliases
- 1810061I13Rik, CLEC2, CLEC2B, PRO1384, QDED721
- Chromosome Location
- Chr. 12: 9986119 - 10001894 on Build GRCh38
- UniGene ID
- Hs.409794
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001099431.1 |
NP_001092901.1
|
3, 4 |
537 |
|
NM_016509.3 |
NP_057593.3
|
4, 5 |
636 |
|
XM_005253382.4 |
XP_005253439.1
|
6, 7 |
1008 |
|
XM_011520685.2 |
XP_011518987.1
|
5, 6 |
917 |
|
XM_011520686.2 |
XP_011518988.1
|
4, 5 |
545 |
|
XM_017019395.1 |
XP_016874884.1
|
5, 6 |
992 |
|
XM_017019396.1 |
XP_016874885.1
|
3, 4 |
344 |
|
GenBank mRNA |
AF124841.1 |
- |
4, 5 |
493 |
|
AY358599.1 |
- |
4, 5 |
617 |
|
BC029554.1 |
- |
5, 6 |
608 |
5269252 |
BX647321.1 |
- |
3, 4 |
328 |
|
View Full Product Details