Gene Transcripts

Gene Symbol
GCNT4
Entrez Gene ID
51301
Gene Name
glucosaminyl (N-acetyl) transferase 4, core 2
Gene Aliases
C2GNT3
Chromosome Location
Chr. 5: 75027464 - 75030899 on Build GRCh38
UniGene ID
Hs.272404
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016591.2 NP_057675.1 1 1643
GenBank mRNA AF132035.1 - 1 1642

View Full Product Details