Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015986.3 | NP_057070.3 | 4 | 696 | |
NR_073118.1 | 3 | 488 | |||
XM_011524879.1 | XP_011523181.1 | 4 | 870 | ||
GenBank mRNA | AF046059.1 | - | 4 | 609 | |
AF120151.1 | - | 314 | |||
AK296238.1 | - | 3 | 399 | ||
AK313315.1 | - | 4 | 685 | ||
BC023567.2 | - | 4 | 618 | 4309046 | |
BU902521.1 | - | 4 | 737 | ||
DQ298450.1 | - | 4 | 575 |