Gene Transcripts

Gene Symbol
ANAPC5
Entrez Gene ID
51433
Gene Name
anaphase promoting complex subunit 5
Gene Aliases
APC5
Chromosome Location
Chr. 12: 121308245 - 121354209 on Build GRCh38
UniGene ID
Hs.7101
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001137559.1 NP_001131031.1 7 718
NM_016237.4 NP_057321.2 7 940
XM_005253900.3 XP_005253957.1 7 942
XM_017019423.1 XP_016874912.1 7 658
XM_017019424.1 XP_016874913.1 7 658
GenBank mRNA AF191339.1 - 7 818
AK025614.1 - 7 718
AK294139.1 - 1 18
AK315451.1 - 7 885
BC001081.2 - 7 873 2823401
BC001950.1 - 7 873 2823401
BC003095.1 - 2 162
BC006301.2 - 7 877 4100632
BC034243.1 - 6 580
BX537687.1 - 4 621

View Full Product Details