Gene Transcripts
- Gene Symbol
- RAB14
- Entrez Gene ID
- 51552
- Gene Name
- RAB14, member RAS oncogene family
- Gene Aliases
- FBP, RAB-14
- Chromosome Location
- Chr. 9: 121178137 - 121202087 on Build GRCh38
- UniGene ID
- Hs.371563
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016322.3 |
NP_057406.2
|
7, 8 |
935 |
|
GenBank mRNA |
AF112206.1 |
- |
7, 8 |
543 |
|
AF152463.1 |
- |
6, 7 |
479 |
|
AF203689.1 |
- |
6, 7 |
485 |
|
AF498949.1 |
- |
6, 7 |
479 |
|
AK023524.1 |
- |
7, 8 |
717 |
|
AK090889.1 |
- |
7, 8 |
734 |
|
AL162081.1 |
- |
7, 8 |
662 |
|
AY553875.1 |
- |
6, 7 |
479 |
|
BC006081.1 |
- |
7, 8 |
706 |
2963119 |
CR457394.1 |
- |
6, 7 |
479 |
|
View Full Product Details