Gene Transcripts

Gene Symbol
RAB6B
Entrez Gene ID
51560
Gene Name
RAB6B, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 3: 133824235 - 133895847 on Build GRCh38
UniGene ID
Hs.715344
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016577.3 NP_057661.3 6 824
XM_011512893.2 XP_011511195.1 6 499
GenBank mRNA AF166492.1 - 6 780
AF498940.1 - 6 443
AK295451.1 - 7 561
AK312378.1 - 6 744
BC002510.2 - 6 793 3050592
BC078662.1 - 6 572 6450986
BT007263.1 - 6 443

View Full Product Details