Gene Transcripts
- Gene Symbol
- METTL13
- Entrez Gene ID
- 51603
- Gene Name
- methyltransferase like 13
- Gene Aliases
- 5630401D24Rik, CGI-01, KIAA0859, feat
- Chromosome Location
- Chr. 1: 171781504 - 171823962 on Build GRCh38
- UniGene ID
- Hs.494705
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001007239.1 |
NP_001007240.1
|
5, 6 |
1358 |
|
NM_014955.2 |
NP_055770.1
|
5, 6 |
1501 |
|
NM_015935.4 |
NP_057019.3
|
5, 6 |
1826 |
|
XM_017001455.1 |
XP_016856944.1
|
4, 5 |
1320 |
|
XM_017001456.1 |
XP_016856945.1
|
4, 5 |
1320 |
|
GenBank mRNA |
AB020666.2 |
- |
5, 6 |
1581 |
|
AB242174.1 |
- |
5, 6 |
1751 |
|
AB242175.1 |
- |
5, 6 |
1739 |
|
AF132936.1 |
- |
5, 6 |
1751 |
|
AK001172.1 |
- |
3, 4 |
513 |
|
AK027552.1 |
- |
5, 6 |
1745 |
|
AK027621.1 |
- |
5, 6 |
1440 |
|
AK027886.1 |
- |
4, 5 |
1297 |
|
AK129561.1 |
- |
4, 5 |
1174 |
|
AK291740.1 |
- |
5, 6 |
1773 |
|
AK304471.1 |
- |
6, 7 |
1607 |
|
AL049669.1 |
- |
5, 6 |
1283 |
|
BC006814.1 |
- |
2, 3 |
376 |
|
BC029083.1 |
- |
5, 6 |
1580 |
4509086 |
View Full Product Details