Gene Transcripts
- Gene Symbol
- ZFR
- Entrez Gene ID
- 51663
- Gene Name
- zinc finger RNA binding protein
- Gene Aliases
- SPG71, ZFR1
- Chromosome Location
- Chr. 5: 32354350 - 32444738 on Build GRCh38
- UniGene ID
- Hs.435231
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016107.3 |
NP_057191.2
|
17, 18 |
2924 |
|
GenBank mRNA |
AF100742.1 |
- |
7, 8 |
919 |
|
AJ314790.1 |
- |
17, 18 |
2880 |
|
AK025481.1 |
- |
5, 6 |
617 |
|
AK123306.1 |
- |
1, 2 |
1320 |
|
AK295297.1 |
- |
4, 5 |
470 |
|
AK307808.1 |
- |
3, 4 |
262 |
|
BC051893.1 |
- |
10, 11 |
1515 |
|
BC062986.1 |
- |
10, 11 |
1440 |
|
BC137084.1 |
- |
17, 18 |
2888 |
9021081 |
BC137085.1 |
- |
17, 18 |
2888 |
9021082 |
View Full Product Details