Gene Transcripts
- Gene Symbol
- DCUN1D1
- Entrez Gene ID
- 54165
- Gene Name
- defective in cullin neddylation 1 domain containing 1
- Gene Aliases
- DCNL1, DCUN1L1, RP42, SCCRO, SCRO, Tes3
- Chromosome Location
- Chr. 3: 182938158 - 182986364 on Build GRCh38
- UniGene ID
- Hs.592789
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001308101.1 |
NP_001295030.1
|
5, 6 |
743 |
|
NM_020640.3 |
NP_065691.2
|
5, 6 |
725 |
|
XM_011512912.1 |
XP_011511214.1
|
5, 6 |
694 |
|
XM_011512914.1 |
XP_011511216.1
|
5, 6 |
733 |
|
XM_011512915.2 |
XP_011511217.1
|
6, 7 |
1255 |
|
XM_011512916.2 |
XP_011511218.1
|
5, 6 |
1183 |
|
XM_017006633.1 |
XP_016862122.1
|
4, 5 |
610 |
|
XM_017006634.1 |
XP_016862123.1
|
5, 6 |
699 |
|
XM_017006635.1 |
XP_016862124.1
|
6, 7 |
1255 |
|
GenBank mRNA |
AF292100.2 |
- |
5, 6 |
642 |
|
AF456425.1 |
- |
5, 6 |
628 |
|
AF456426.1 |
- |
5, 6 |
628 |
|
AK025764.1 |
- |
3, 4 |
288 |
|
AK056335.1 |
- |
5, 6 |
658 |
|
AK297328.1 |
- |
6, 7 |
542 |
|
AK314480.1 |
- |
5, 6 |
642 |
|
BC009478.2 |
- |
5, 6 |
647 |
3939758 |
BC013163.1 |
- |
4, 5 |
598 |
|
BI828039.1 |
- |
5, 6 |
683 |
|
View Full Product Details