Gene Transcripts
- Gene Symbol
- SNTG1
- Entrez Gene ID
- 54212
- Gene Name
- syntrophin gamma 1
- Gene Aliases
- G1SYN, SYN4
- Chromosome Location
- Chr. 8: 49909726 - 50796656 on Build GRCh38
- UniGene ID
- Hs.584914
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001287813.2 |
NP_001274742.1
|
11, 12 |
982 |
|
NM_001287814.2 |
NP_001274743.1
|
9, 10 |
1134 |
|
NM_001321773.1 |
NP_001308702.1
|
9, 10 |
1134 |
|
NM_001321775.1 |
NP_001308704.1
|
11, 12 |
982 |
|
NM_001321776.1 |
NP_001308705.1
|
10, 11 |
1209 |
|
NM_001321777.1 |
NP_001308706.1
|
9, 10 |
1134 |
|
NM_001321778.1 |
NP_001308707.1
|
10, 11 |
940 |
|
NM_018967.4 |
NP_061840.1
|
10, 11 |
1209 |
|
NR_135794.1 |
|
10, 11 |
1189 |
|
NR_135795.1 |
|
10, 11 |
1209 |
|
NR_135796.1 |
|
10, 11 |
1209 |
|
NR_135797.1 |
|
10, 11 |
1189 |
|
NR_135798.1 |
|
9, 10 |
1134 |
|
XM_011517548.2 |
XP_011515850.1
|
9, 10 |
1319 |
|
XM_017013579.1 |
XP_016869068.1
|
12, 13 |
4138 |
|
XM_017013580.1 |
XP_016869069.1
|
10, 11 |
732 |
|
XM_017013581.1 |
XP_016869070.1
|
10, 11 |
1454 |
|
XM_017013582.1 |
XP_016869071.1
|
10, 11 |
1454 |
|
GenBank mRNA |
AJ003030.1 |
- |
9, 10 |
770 |
|
AK314084.1 |
- |
11, 12 |
919 |
|
AL161971.1 |
- |
10, 11 |
1104 |
|
BC075072.2 |
- |
8, 9 |
574 |
30915592 |
BC104829.1 |
- |
9, 10 |
656 |
8143832 |
BC143695.1 |
- |
9, 10 |
656 |
|
View Full Product Details