Gene Transcripts
- Gene Symbol
- EPDR1
- Entrez Gene ID
- 54749
- Gene Name
- ependymin related 1
- Gene Aliases
- EPDR, MERP-1, MERP1, UCC1
- Chromosome Location
- Chr. 7: 37920561 - 37951941 on Build GRCh38
- UniGene ID
- Hs.563491
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001242948.1 |
NP_001229877.1
|
2, 3 |
330 |
|
NM_017549.4 |
NP_060019.2
|
2, 3 |
845 |
|
GenBank mRNA |
AJ250475.2 |
- |
2, 3 |
318 |
|
AK290885.1 |
- |
2, 3 |
539 |
|
AK309180.1 |
- |
2, 3 |
317 |
|
AY027862.2 |
- |
2, 3 |
845 |
|
BC000686.2 |
- |
2, 3 |
589 |
|
BC018299.1 |
- |
2, 3 |
737 |
3461888 |
DA938402.1 |
- |
3, 4 |
358 |
|
DB167672.1 |
- |
2, 3 |
331 |
|
View Full Product Details