Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001312691.1 | NP_001299620.1 | 7 | 849 | |
NM_001312692.1 | NP_001299621.1 | 5 | 603 | ||
NM_017646.5 | NP_060116.2 | 7 | 849 | ||
NR_132401.1 | 7 | 845 | |||
NR_132402.1 | 6 | 703 | |||
NR_132403.1 | 6 | 699 | |||
NR_132404.1 | 5, 6 | 699 | |||
NR_132405.1 | 5, 6 | 695 | |||
NR_132407.1 | 4 | 463 | |||
NR_132408.1 | 4 | 459 | |||
NR_132409.1 | 3 | 320 | |||
NR_132415.1 | 7 | 936 | |||
XM_005270954.2 | XP_005271011.1 | 7 | 1085 | ||
XM_006710706.1 | XP_006710769.1 | 7 | 917 | ||
GenBank mRNA | AF074918.1 | - | 7 | 930 | |
AK000068.1 | - | 5 | 466 | ||
AK074222.1 | - | 5 | 466 | ||
AK223478.1 | - | 7 | 842 | ||
AK307910.1 | - | 7 | 834 | ||
AY052768.1 | - | 7 | 811 | ||
AY303390.1 | - | 7 | 843 | ||
AY702933.1 | - | 7 | 772 | ||
AY702934.1 | - | 7 | 768 | ||
AY702935.1 | - | 6 | 619 | ||
AY702936.1 | - | 6 | 626 | ||
AY702937.1 | - | 6 | 622 | ||
AY702938.1 | - | 5, 6 | 622 | ||
AY702939.1 | - | 5, 6 | 618 | ||
AY702941.1 | - | 4 | 386 | ||
AY702942.1 | - | 4 | 382 | ||
AY702943.1 | - | 3 | 243 | ||
BC010741.1 | - | 5 | 565 | ||
BC107569.1 | - | 7 | 848 | 4594069 | |
BC128155.1 | - | 7 | 843 | 40112695 | |
BC128156.1 | - | 7 | 930 | 40112698 | |
BE294565.1 | - | 4 | 465 | ||
CR457226.1 | - | 4 | 400 |