Gene Transcripts

Gene Symbol
PALMD
Entrez Gene ID
54873
Gene Name
palmdelphin
Gene Aliases
C1orf11, PALML
Chromosome Location
Chr. 1: 99645875 - 99694541 on Build GRCh38
UniGene ID
Hs.483993
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017734.4 NP_060204.1 6 907
GenBank mRNA AF262379.1 - 6 838
AJ312214.1 - 6 749
AK000278.1 - 6 669
AL390971.1 - 6 837
AL390972.1 - 6 346
AL390973.1 - 6 633
BC012570.1 - 6 722 4245261
BX648213.1 - 3 3622
KU178657.1 - 6 464
KU178658.1 - 6 464

View Full Product Details