Gene Transcripts

Gene Symbol
C20orf27
Entrez Gene ID
54976
Gene Name
chromosome 20 open reading frame 27
Gene Aliases
Chromosome Location
Chr. 20: 3753499 - 3768388 on Build GRCh38
UniGene ID
Hs.274422
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039140.2 NP_001034229.1 6 897
NM_001258429.1 NP_001245358.1 6 822
NM_001258430.1 NP_001245359.1 6 1276
NR_047675.1 6 966
XM_011529266.2 XP_011527568.1 5 754
GenBank mRNA AK000557.1 - 6 761
AK024220.1 - 6 761
AK097645.1 - 6 1404
AK124341.1 - 4 2014
AK125202.1 - 3 1968
AK225109.1 - 6 761
AK290955.1 - 6 966
BC012196.1 - 6 748 2822218
BC024036.1 - 6 759 3510659

View Full Product Details