Gene Transcripts

Gene Symbol
PLEKHJ1
Entrez Gene ID
55111
Gene Name
pleckstrin homology domain containing J1
Gene Aliases
GNRPX
Chromosome Location
Chr. 19: 2229952 - 2236724 on Build GRCh38
UniGene ID
Hs.501353
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300836.1 NP_001287765.1 4 390
NM_018049.2 NP_060519.1 4 390
XM_017026924.1 XP_016882413.1 4 713
XM_017026925.1 XP_016882414.1 4 704
XM_017026926.1 XP_016882415.1 4 713
XM_017026927.1 XP_016882416.1 4 704
XM_017026928.1 XP_016882417.1 4 704
GenBank mRNA AK001159.1 - 4 366
AK092742.1 - 2 1915
AK097723.1 - 5 1397
AK098725.1 - 4 480
BC003084.2 - 4 311 2988404
BC008966.2 - 4 337 2961278
CN365243.1 - 4 309
CR457253.1 - 4 285
HY085955.1 - 4 380
HY101097.1 - 4 393

View Full Product Details