Gene Transcripts

Gene Symbol
CRTAC1
Entrez Gene ID
55118
Gene Name
cartilage acidic protein 1
Gene Aliases
ASPIC, ASPIC1, CEP-68
Chromosome Location
Chr. 10: 97854711 - 98030828 on Build GRCh38
UniGene ID
Hs.500736
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206528.2 NP_001193457.1 5 935
NM_018058.6 NP_060528.3 5 935
XM_005269938.4 XP_005269995.1 5 728
XM_011539917.1 XP_011538219.1 5 728
XM_017016366.1 XP_016871855.1 5 728
XM_017016367.1 XP_016871856.1 5 728
GenBank mRNA AJ276171.1 - 5 728
AJ279016.1 - 4 555
AJ421515.1 - 4 555
AK001182.1 - 3 267
AK057190.1 - 5 935
AK292084.1 - 5 697
BC034245.1 - 3 197 4178363

View Full Product Details