Gene Transcripts

Gene Symbol
LRRC8D
Entrez Gene ID
55144
Gene Name
leucine rich repeat containing 8 family member D
Gene Aliases
LRRC5
Chromosome Location
Chr. 1: 89821014 - 89936432 on Build GRCh38
UniGene ID
Hs.482087
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134479.1 NP_001127951.1 3 444
NM_018103.4 NP_060573.2 3 431
XM_011541689.1 XP_011539991.1 4 515
XM_017001599.1 XP_016857088.1 3 786
XM_017001600.1 XP_016857089.1 3 241
XM_017001601.1 XP_016857090.1 3 1132
XM_017001602.1 XP_016857091.1 3 485
GenBank mRNA AK092238.1 - 3 407
BC009486.1 - 3 376
BC024159.2 - 3 398 3958069
BP283613.1 - 3 431
DA126452.1 - 3 427
DB097950.1 - 4 509

View Full Product Details