Gene Transcripts

Gene Symbol
VPS13D
Entrez Gene ID
55187
Gene Name
vacuolar protein sorting 13 homolog D
Gene Aliases
Chromosome Location
Chr. 1: 12230039 - 12512047 on Build GRCh38
UniGene ID
Hs.439381
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015378.3 NP_056193.2 49 10112
NM_018156.3 NP_060626.2 48 10037
GenBank mRNA AB007922.3 - 31 6419
AJ608774.1 - 49 10084
AJ608775.1 - 48 10009
AK057675.1 - 7 1081
AK125118.1 - 23 3957
AL833079.1 - 4 723
AL833320.1 - 438
BC028115.1 - 7 1028
BX641035.1 - 23 3601

View Full Product Details