Gene Transcripts
- Gene Symbol
- TMEM40
- Entrez Gene ID
- 55287
- Gene Name
- transmembrane protein 40
- Gene Aliases
- Chromosome Location
- Chr. 3: 12733530 - 12769457 on Build GRCh38
- UniGene ID
- Hs.475502
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001284406.1 |
NP_001271335.1
|
7, 8 |
649 |
|
NM_001284407.1 |
NP_001271336.1
|
6, 7 |
471 |
|
NM_001284408.1 |
NP_001271337.1
|
5, 6 |
333 |
|
NM_018306.3 |
NP_060776.2
|
7, 8 |
561 |
|
XM_011533937.2 |
XP_011532239.1
|
8, 9 |
816 |
|
GenBank mRNA |
AK001898.1 |
- |
6, 7 |
403 |
|
AK092470.1 |
- |
7, 8 |
561 |
|
AK301984.1 |
- |
7, 8 |
649 |
|
BC020658.1 |
- |
6, 7 |
421 |
4768606 |
BG335433.1 |
- |
5, 6 |
272 |
|
View Full Product Details