Gene Transcripts

Gene Symbol
SLC22A15
Entrez Gene ID
55356
Gene Name
solute carrier family 22 member 15
Gene Aliases
FLIPT1, PRO34686
Chromosome Location
Chr. 1: 115976498 - 116070054 on Build GRCh38
UniGene ID
Hs.125482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018420.2 NP_060890.2 6 876
XM_005271004.3 XP_005271061.1 6 837
GenBank mRNA AK125072.1 - 4 444
AY145501.1 - 6 746
BC026358.1 - 6 875 4814675
KU178698.1 - 6 710
KU178700.1 - 6 710
KU178703.1 - 5 497

View Full Product Details