Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001206864.1 | NP_001193793.1 | 6 | 717 | |
NM_018571.5 | NP_061041.2 | 6 | 717 | ||
XM_005246669.1 | XP_005246726.1 | 6 | 671 | ||
XM_017004438.1 | XP_016859927.1 | 5 | 552 | ||
GenBank mRNA | AB038950.1 | - | 6 | 593 | |
AF116618.2 | - | 2 | 154 | ||
AK024720.1 | - | 2 | 71 | ||
AK027637.1 | - | 6 | 717 | ||
AK290457.1 | - | 7 | 581 | ||
AY093697.1 | - | 6 | 605 | ||
BC008302.1 | - | 6 | 619 | 3546243 | |
BC090871.1 | - | 6 | 590 | 30530122 |