Gene Transcripts

Gene Symbol
PIGV
Entrez Gene ID
55650
Gene Name
phosphatidylinositol glycan anchor biosynthesis class V
Gene Aliases
GPI-MT-II, HPMRS1, PIG-V
Chromosome Location
Chr. 1: 26787963 - 26798403 on Build GRCh38
UniGene ID
Hs.259605
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001202554.1 NP_001189483.1 3 1768
NM_017837.3 NP_060307.2 3 1442
GenBank mRNA AK000484.1 - 3 1217
AK056539.1 - 3 1761
AK225521.1 - 3 1217
AK301626.1 - 4 747
BC013568.1 - 3 1410 3890907

View Full Product Details