Gene Transcripts

Gene Symbol
CCAR1
Entrez Gene ID
55749
Gene Name
cell division cycle and apoptosis regulator 1
Gene Aliases
Chromosome Location
Chr. 10: 68721144 - 68792377 on Build GRCh38
UniGene ID
Hs.49853
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282959.1 NP_001269888.1 14 2046
NM_001282960.1 NP_001269889.1 14 2040
NM_018237.3 NP_060707.2 15 2085
NR_104262.1 14 1880
GenBank mRNA AF465616.1 - 15 2015
AK000741.1 - 10 1399
AK001701.1 - 15 1993
AK023438.1 - 15 2015
AK122871.1 - 13 1860
AK292649.1 - 15 1990
AK298004.1 - 14 1965
AK299307.1 - 14 1945
AK307550.1 - 10 1405
AK307671.1 - 13 1909
AK307704.1 - 15 1990
AY249140.1 - 15 1993
BC015475.1 - 5 624
BC026036.2 - 14 1947
BC089420.1 - 14 1907
BC108682.1 - 15 1963
BC130626.1 - 15 1974 40146657
BC132725.1 - 15 1974 40146747
BC144118.1 - 14 1769
BX647943.1 - 13 1820

View Full Product Details