Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008744.1 | NP_001008744.1 | 10 | 1298 | |
NM_018319.3 | NP_060789.2 | 11 | 1521 | ||
XM_005267847.2 | XP_005267904.1 | 11 | 2195 | ||
XM_005267848.2 | XP_005267905.1 | 10 | 1539 | ||
XM_006720197.3 | XP_006720260.1 | 11 | 1762 | ||
XM_006720198.3 | XP_006720261.1 | 10 | 1417 | ||
XM_006720199.1 | XP_006720262.1 | 10 | 903 | ||
XM_011536942.2 | XP_011535244.1 | 10 | 1938 | ||
XM_011536943.2 | XP_011535245.1 | 10 | 1331 | ||
XM_011536944.1 | XP_011535246.1 | 11 | 1522 | ||
XM_017021439.1 | XP_016876928.1 | 11 | 1429 | ||
XM_017021440.1 | XP_016876929.1 | 10 | 1321 | ||
XM_017021441.1 | XP_016876930.1 | 10 | 1510 | ||
XM_017021442.1 | XP_016876931.1 | 8 | 1147 | ||
XM_017021443.1 | XP_016876932.1 | 8 | 646 | ||
XM_017021444.1 | XP_016876933.1 | 8 | 647 | ||
XM_017021445.1 | XP_016876934.1 | 8 | 1147 | ||
XM_017021446.1 | XP_016876935.1 | 9 | 953 | ||
GenBank mRNA | AF182002.1 | - | 9 | 1207 | |
AK001952.1 | - | 10 | 1262 | ||
AK093235.1 | - | 12 | 2329 | ||
AK315549.1 | - | 10 | 1246 | ||
AL832288.1 | - | 2 | 240 | ||
BC006083.1 | - | 13 | 1685 | ||
BC015474.1 | - | 11 | 1472 | 3900062 | |
BC107877.1 | - | 11 | 1525 | 6597752 | |
BX161451.1 | - | 10 | 1433 |