Gene Transcripts

Gene Symbol
UBAP2
Entrez Gene ID
55833
Gene Name
ubiquitin associated protein 2
Gene Aliases
UBAP-2
Chromosome Location
Chr. 9: 33921693 - 34048949 on Build GRCh38
UniGene ID
Hs.493739
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282529.1 NP_001269458.1 11 1496
NM_018449.3 NP_060919.3 15 1810
GenBank mRNA AB040924.1 - 3 612
AK025801.1 - 4 700
AK026088.1 - 7 978
AK074884.1 - 2 169
AK294952.1 - 15 1728
AK299183.1 - 9 1218
AK302335.1 - 13 1631
AK302754.1 - 14 1678
AK303493.1 - 11 1434
AY358682.1 - 3 1020
BC111762.1 - 9 1180

View Full Product Details