Gene Transcripts

Gene Symbol
KCNQ5
Entrez Gene ID
56479
Gene Name
potassium voltage-gated channel subfamily Q member 5
Gene Aliases
Kv7.5
Chromosome Location
Chr. 6: 72621843 - 73198851 on Build GRCh38
UniGene ID
Hs.445324
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160130.1 NP_001153602.1 6 1326
NM_001160132.1 NP_001153604.1 6 1326
NM_001160133.1 NP_001153605.1 6 1326
NM_001160134.1 NP_001153606.1 6 1326
NM_019842.3 NP_062816.2 6 1326
XM_017011058.1 XP_016866547.1 6 1326
GenBank mRNA AF202977.1 - 6 874
AF249278.1 - 6 1088
AF263835.1 - 6 766
AK299550.1 - 6 926
BC050689.1 - 6 1063
BC117359.1 - 6 997 40125910
BC143554.1 - 6 997 9052065
BC143555.1 - 5 870

View Full Product Details