Gene Transcripts
- Gene Symbol
- NIT2
- Entrez Gene ID
- 56954
- Gene Name
- nitrilase family member 2
- Gene Aliases
- HEL-S-8a
- Chromosome Location
- Chr. 3: 100334718 - 100355634 on Build GRCh38
- UniGene ID
- Hs.439152
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020202.4 |
NP_064587.1
|
2, 3 |
207 |
|
GenBank mRNA |
AF260334.1 |
- |
2, 3 |
206 |
|
AF284574.1 |
- |
2, 3 |
154 |
|
AK225979.1 |
- |
2, 3 |
183 |
|
AK294010.1 |
- |
2, 3 |
168 |
|
AK295835.1 |
- |
2, 3 |
168 |
|
AK313704.1 |
- |
2, 3 |
149 |
|
BC020620.1 |
- |
2, 3 |
164 |
4775062 |
BC107890.1 |
- |
2, 3 |
195 |
6648910 |
DB466042.1 |
- |
2, 3 |
207 |
|
FJ224326.1 |
- |
2, 3 |
154 |
|
View Full Product Details