Gene Transcripts

Gene Symbol
ADPRM
Entrez Gene ID
56985
Gene Name
ADP-ribose/CDP-alcohol diphosphatase, manganese dependent
Gene Aliases
C17orf48, MDS006, NBLA03831
Chromosome Location
Chr. 17: 10697610 - 10711558 on Build GRCh38
UniGene ID
Hs.47668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020233.4 NP_064618.3 2 244
XM_005256738.2 XP_005256795.1 2 244
GenBank mRNA AB073393.1 - 2 215
AF168715.1 - 1 208
AK292629.1 - 2 210
BC001294.2 - 1 199 3452502
BC070155.1 - 2 239 4656998
DA305828.1 - 2 244
KF880964.1 - 1 169

View Full Product Details