Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021035.2 | NP_066363.1 | 3 | 790 | |
XM_006723836.1 | XP_006723899.1 | 3 | 620 | ||
XM_011528924.1 | XP_011527226.1 | 3 | 815 | ||
GenBank mRNA | AB037825.2 | - | 3 | 608 | |
AK002139.1 | - | 3 | 703 | ||
AK096594.1 | - | 3 | 997 | ||
AK307392.1 | - | 3 | 994 | ||
BC014041.2 | - | 2 | 606 |