Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017519.2 | NP_059989.2 | 4 | 1955 | |
NM_020732.3 | NP_065783.3 | 5 | 1994 | ||
XM_005267069.4 | XP_005267126.2 | 4 | 2030 | ||
XM_011535984.2 | XP_011534286.2 | 5 | 2070 | ||
XM_017011103.1 | XP_016866592.1 | 5 | 2070 | ||
XM_017011104.1 | XP_016866593.1 | 5 | 2069 | ||
XM_017011105.1 | XP_016866594.1 | 5 | 2070 | ||
XM_017011106.1 | XP_016866595.1 | 5 | 2069 | ||
XM_017011107.1 | XP_016866596.1 | 5 | 2070 | ||
XM_017011108.1 | XP_016866597.1 | 5 | 2077 | ||
GenBank mRNA | AF253515.1 | - | 4 | 1149 | |
AF468300.1 | - | 5 | 524 | ||
AF521671.1 | - | 4 | 1742 | ||
AK025945.1 | - | 2 | 221 | ||
AK074180.1 | - | 1 | 63 |