Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020772.2 | NP_065823.1 | 2 | 1398 | |
XM_017024896.1 | XP_016880385.1 | 2 | 2696 | ||
GenBank mRNA | AB037742.1 | - | 2 | 1368 | |
AJ493465.1 | - | 2 | 1309 | ||
AK293075.1 | - | 2 | 1398 | ||
AY232289.1 | - | 2 | 1377 | ||
BC108307.1 | - | 2 | 1355 | 6502841 | |
BC129990.1 | - | 2 | 1313 | 40128910 |