Gene Transcripts

Gene Symbol
WDFY1
Entrez Gene ID
57590
Gene Name
WD repeat and FYVE domain containing 1
Gene Aliases
FENS-1, WDF1, ZFYVE17
Chromosome Location
Chr. 2: 223875343 - 223945387 on Build GRCh38
UniGene ID
Hs.368359
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020830.4 NP_065881.1 11, 12 1266
GenBank mRNA AB037856.1 - 11, 12 1179
AF411977.1 - 11, 12 1163
AJ310568.1 - 11, 12 1253
AK022888.1 - 11, 12 1192
AK023415.1 - 10, 11 1060
BC040525.1 - 11, 12 1178 5285567
BC065934.1 - 11, 12 1210 6164567

View Full Product Details