Gene Transcripts
- Gene Symbol
- PLEKHA4
- Entrez Gene ID
- 57664
- Gene Name
- pleckstrin homology domain containing A4
- Gene Aliases
- PEPP1
- Chromosome Location
- Chr. 19: 48837092 - 48868627 on Build GRCh38
- UniGene ID
- Hs.9469
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001161354.1 |
NP_001154826.1
|
16, 17 |
2326 |
|
NM_020904.2 |
NP_065955.2
|
19, 20 |
2622 |
|
XM_005259107.2 |
XP_005259164.1
|
18, 19 |
2542 |
|
XM_006723301.2 |
XP_006723364.1
|
17, 18 |
2483 |
|
XM_011527157.1 |
XP_011525459.1
|
19, 20 |
2629 |
|
XM_011527158.2 |
XP_011525460.1
|
18, 19 |
2468 |
|
XM_011527159.1 |
XP_011525461.1
|
19, 20 |
2642 |
|
XM_011527160.1 |
XP_011525462.1
|
18, 19 |
2570 |
|
XM_011527162.2 |
XP_011525464.1
|
12, 13 |
1238 |
|
XM_017027037.1 |
XP_016882526.1
|
16, 17 |
1906 |
|
GenBank mRNA |
AB208908.1 |
- |
17, 18 |
4114 |
|
AY007233.1 |
- |
19, 20 |
2596 |
|
BC024157.2 |
- |
19, 20 |
2602 |
3953478 |
BC033832.1 |
- |
11, 12 |
1072 |
|
BC064601.1 |
- |
16, 17 |
2153 |
5785991 |
View Full Product Details