Gene Transcripts

Gene Symbol
EPB41L5
Entrez Gene ID
57669
Gene Name
erythrocyte membrane protein band 4.1 like 5
Gene Aliases
BE37, YMO1, YRT
Chromosome Location
Chr. 2: 120013028 - 120179121 on Build GRCh38
UniGene ID
Hs.369232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184937.1 NP_001171866.1 5, 6 605
NM_001184938.2 NP_001171867.1 5, 6 895
NM_001184939.1 NP_001171868.1 5, 6 605
NM_020909.3 NP_065960.2 5, 6 605
NR_135920.1 5, 6 605
XM_006712651.1 XP_006712714.1 5, 6 585
XM_006712652.1 XP_006712715.1 5, 6 900
XM_006712653.1 XP_006712716.1 5, 6 605
XM_011511529.2 XP_011509831.1 5, 6 585
XM_017004567.1 XP_016860056.1 5, 6 441
XM_017004568.1 XP_016860057.1 5, 6 605
GenBank mRNA AK023019.1 - 5, 6 556
AK290895.1 - 5, 6 515
BC032822.2 - 5, 6 895 4827274
BC054508.1 - 5, 6 540 5502663

View Full Product Details