Gene Transcripts

Gene Symbol
PLEKHB1
Entrez Gene ID
58473
Gene Name
pleckstrin homology domain containing B1
Gene Aliases
KPL1, PHR1, PHRET1
Chromosome Location
Chr. 11: 73646178 - 73662819 on Build GRCh38
UniGene ID
Hs.445489
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130033.1 NP_001123505.1 3 591
NM_001130034.1 NP_001123506.1 3 332
NM_001130035.1 NP_001123507.1 3 332
NM_001130036.1 NP_001123508.1 2 464
NM_021200.2 NP_067023.1 3 591
XM_011545192.2 XP_011543494.1 3 1165
XM_011545193.2 XP_011543495.1 3 1165
GenBank mRNA AB209716.1 - 3 244
AF081583.1 - 2 108
AF093249.1 - 3 216
AF100612.1 - 3 216
AF101054.1 - 3 587
AK289620.1 - 3 244
AK289909.1 - 3 210
AK290109.1 - 2 464
AK296033.1 - 3 226
AK301299.1 - 3 227
AK314748.1 - 3 188
BC008075.2 - 3 270 3050369
DC305763.1 - 3 332
U89715.1 - 3 587

View Full Product Details